siRNA Details
| Gene Symbol: Spats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139140.1 | NP_631879.1 | 4 | 434 | ||
| GenBank mRNA | AB079132.1 | - | 4 | 434 | ||
| AK048720.1 | - | 4 | 476 | |||
| AK135838.1 | - | 4 | 492 | |||
| AK159335.1 | - | 4 | 528 | |||
| AK167788.1 | - | 3 | 111 | |||
| AK171266.1 | - | 3 | 189 | |||
| BC023432.1 | - | 2 | 84 | 5029821 | ||
| BC072579.1 | - | 4 | 701 | |||

Spats2
Genomic Map