siRNA Details
| Gene Symbol: Dppa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139218.1 | NP_631964.1 | 4 | 563 | ||
| GenBank mRNA | AF490347.1 | - | 4 | 568 | ||
| AK135769.1 | - | 4 | 568 | |||
| AK136263.1 | - | 4 | 568 | |||
| AY082485.1 | - | 4 | 563 | |||
| AY134859.1 | - | 4 | 575 | |||
| BC099433.1 | - | 4 | 541 | 30933569 | ||
| BC100331.1 | - | 4 | 538 | 30934920 | ||
| BC107340.1 | - | 4 | 495 | 40055218 | ||
| BC107341.1 | - | 4 | 495 | 40055220 | ||

Dppa3
Genomic Map