siRNA Details
| Gene Symbol: Kazn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144531.3 | NP_653114.3 | 14 | 3204 | ||
| GenBank mRNA | AK032460.1 | - | 12 | 2443 | ||
| AK173090.1 | - | 14 | 3204 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kazn| Gene Symbol: Kazn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144531.3 | NP_653114.3 | 14 | 3204 | ||
| GenBank mRNA | AK032460.1 | - | 12 | 2443 | ||
| AK173090.1 | - | 14 | 3204 | |||