siRNA Details
| Gene Symbol: Mettl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144877.1 | NP_659126.1 | 8 | 2151 | ||
| GenBank mRNA | AK084750.1 | - | 8 | 1846 | ||
| AK146751.1 | - | 2349 | ||||
| AK168959.1 | - | 8 | 2358 | |||
| AK169319.1 | - | 2350 | ||||
| AK173044.1 | - | 2304 | ||||
| BC014872.1 | - | 8 | 2151 | 3602469 | ||

Mettl13
Genomic Map