siRNA Details
| Gene Symbol: Crtac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145123.4 | NP_660105.3 | 2 | 316 | ||
| GenBank mRNA | AJ421516.1 | - | 2 | 166 | ||
| AK032328.1 | - | 2 | 181 | |||
| AK049801.1 | - | 2 | 287 | |||
| AK083221.1 | - | 2 | 319 | |||
| AK149020.1 | - | 2 | 322 | |||
| AK149069.1 | - | 2 | 317 | |||

Crtac1
Genomic Map