siRNA Details
| Gene Symbol: Crtac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145123.4 | NP_660105.3 | 3 | 463 | ||
| GenBank mRNA | AJ421516.1 | - | 3 | 313 | ||
| AK032328.1 | - | 3 | 328 | |||
| AK049801.1 | - | 3 | 434 | |||
| AK083221.1 | - | 3 | 466 | |||
| AK149020.1 | - | 3 | 469 | |||
| AK149069.1 | - | 3 | 464 | |||

Crtac1
Genomic Map