siRNA Details
| Gene Symbol: Prkag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145401.2 | NP_663376.2 | 2, 3 | 697 | ||
| GenBank mRNA | AK154296.1 | - | 2, 3 | 698 | ||
| BC015283.1 | - | 2, 3 | 485 | 4238045 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkag2| Gene Symbol: Prkag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145401.2 | NP_663376.2 | 2, 3 | 697 | ||
| GenBank mRNA | AK154296.1 | - | 2, 3 | 698 | ||
| BC015283.1 | - | 2, 3 | 485 | 4238045 | ||