siRNA Details
| Gene Symbol: Zfp263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148924.3 | NP_683726.2 | 2, 3 | 851 | ||
| GenBank mRNA | AF499776.1 | - | 2, 3 | 819 | ||
| AK004765.2 | - | 2, 3 | 848 | |||
| AK159455.1 | - | 2, 3 | 839 | |||
| AK182706.1 | - | 2, 3 | 205 | |||
| BC011505.1 | - | 3, 4 | 1377 | |||
| BC056222.1 | - | 2, 3 | 672 | 30009326 | ||
| BC060162.1 | - | 1, 2 | 1784 | |||

Zfp263
Genomic Map