siRNA Details
| Gene Symbol: Osbpl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148958.2 | NP_683761.1 | 6 | 1086 | ||
| GenBank mRNA | AK017034.1 | - | 8 | 1417 | ||
| AK037617.1 | - | 5 | 1228 | |||
| BC027256.1 | - | 6 | 1086 | 2650570 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osbpl10| Gene Symbol: Osbpl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148958.2 | NP_683761.1 | 6 | 1086 | ||
| GenBank mRNA | AK017034.1 | - | 8 | 1417 | ||
| AK037617.1 | - | 5 | 1228 | |||
| BC027256.1 | - | 6 | 1086 | 2650570 | ||