siRNA Details
| Gene Symbol: Cdc5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152810.2 | NP_690023.1 | 2 | 344 | ||
| GenBank mRNA | AK146892.1 | - | 2 | 164 | ||
| AK150562.1 | - | 2 | 161 | |||
| AK150861.1 | - | 2 | 161 | |||
| AK172950.1 | - | 2 | 164 | |||
| BC003893.1 | - | 2 | 169 | 3593535 | ||
| BC031480.1 | - | 2 | 344 | 3498853 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053527, BC161839, BC087007, AF000578.

Cdc5l
Genomic Map