siRNA Details
| Gene Symbol: Oplah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153122.2 | NP_694762.1 | 3 | 333 | ||
| GenBank mRNA | AK158391.1 | - | 3 | 375 | ||
| BC034522.1 | - | 3 | 333 | 4236444 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oplah| Gene Symbol: Oplah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153122.2 | NP_694762.1 | 3 | 333 | ||
| GenBank mRNA | AK158391.1 | - | 3 | 375 | ||
| BC034522.1 | - | 3 | 333 | 4236444 | ||