siRNA Details
| Gene Symbol: Lrrc56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172064.1 | NP_001165535.1 | 8 | 1353 | ||
| NM_001172065.1 | NP_001165536.1 | 8 | 1353 | |||
| NM_153777.2 | NP_722472.1 | 8 | 939 | |||
| GenBank mRNA | AK017603.1 | - | 8 | 1327 | ||
| AK220237.1 | - | 8 | 1307 | |||
| BC027807.1 | - | 8 | 949 | 5324022 | ||

Lrrc56
Genomic Map