siRNA Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170591.1 | NP_733479.1 | 15 | 2207 | ||
| GenBank mRNA | AK129134.1 | - | 14 | 3629 | ||
| AK139895.1 | - | 15 | 2337 | |||
| AK146477.1 | - | 15 | 2207 | |||
| BC026743.1 | - | 15 | 2238 | 3964978 | ||

Nupl1
Genomic Map