siRNA Details
| Gene Symbol: Rap2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172413.2 | NP_766001.1 | 2 | 503 | ||
| GenBank mRNA | AK087971.1 | - | 2 | 450 | ||
| AK146057.1 | - | 2 | 482 | |||
| AK146956.1 | - | 2 | 545 | |||
| AK147015.1 | - | 2 | 545 | |||
| AK214618.1 | - | 2 | 211 | |||
| BC027363.1 | - | 2 | 467 | 4951566 | ||
| BC050056.1 | - | 2 | 465 | 30016070 | ||
| BC064814.1 | - | 2 | 499 | 6513822 | ||
| BC094890.1 | - | 2 | 503 | 30649398 | ||

Rap2c
Genomic Map