siRNA Details
| Gene Symbol: HMGN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006353.2 | NP_006344.1 | 2 | 719 | ||
| XM_017010171.1 | XP_016865660.1 | 2 | 778 | |||
| XM_017010172.1 | XP_016865661.1 | 3 | 842 | |||
| XM_017010173.1 | XP_016865662.1 | 4 | 880 | |||
| GenBank mRNA | AU143224.1 | - | 2 | 719 | ||
| BC001282.1 | - | 2 | 663 | 3452950 | ||
| BC035998.1 | - | 1 | 142 | |||
| U90549.1 | - | 1 | 1172 | |||

HMGN4
Genomic Map