siRNA Details
| Gene Symbol: Asxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270988.1 | NP_001257917.1 | 3 | 533 | ||
| NM_172421.5 | NP_766009.2 | 3 | 533 | |||
| GenBank mRNA | AK016141.1 | - | 2 | 145 | ||
| AK036839.1 | - | 3 | 490 | |||
| AK080679.1 | - | 3 | 507 | |||
| AK142135.1 | - | 3 | 479 | |||
| AK148015.1 | - | 3 | 537 | |||
| BC141140.1 | - | 3 | 367 | 9055752 | ||

Asxl2
Genomic Map