siRNA Details
| Gene Symbol: Asxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270988.1 | NP_001257917.1 | 11 | 1541 | ||
| NM_172421.5 | NP_766009.2 | 10 | 1397 | |||
| GenBank mRNA | AK016141.1 | - | 10 | 1103 | ||
| AK035679.1 | - | 5 | 1969 | |||
| AK036839.1 | - | 10 | 1354 | |||
| AK122540.1 | - | 2 | 170 | |||
| AK148015.1 | - | 10 | 1402 | |||
| AK162455.1 | - | 2 | 138 | |||
| BC141140.1 | - | 11 | 1375 | 9055752 | ||

Asxl2
Genomic Map