siRNA Details
| Gene Symbol: Fndc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173182.2 | NP_775274.2 | 3 | 262 | ||
| GenBank mRNA | AB098596.1 | - | 3 | 200 | ||
| AK045099.1 | - | 3 | 357 | |||
| AK147346.1 | - | 4 | 273 | |||
| AK220297.1 | - | 3 | 262 | |||
| BC067389.1 | - | 3 | 268 | 30431862 | ||
| DQ192037.1 | - | 2 | 168 | |||

Fndc3b
Genomic Map