siRNA Details
| Gene Symbol: XCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024644.1 | NP_001019815.1 | 2 | 617 | ||
| NM_005283.2 | NP_005274.1 | 3 | 739 | |||
| XM_017006196.1 | XP_016861685.1 | 4 | 1757 | |||
| XM_017006197.1 | XP_016861686.1 | 6 | 2009 | |||
| XM_017006198.1 | XP_016861687.1 | 5 | 1822 | |||
| XM_017006199.1 | XP_016861688.1 | 4 | 819 | |||
| XM_017006200.1 | XP_016861689.1 | 3 | 1677 | |||
| XM_017006201.1 | XP_016861690.1 | 2 | 1526 | |||
| GenBank mRNA | AK314277.1 | - | 2 | 870 | ||
| BC069075.1 | - | 1 | 515 | 7216889 | ||
| BC069589.1 | - | 1 | 515 | 7262280 | ||
| BC095533.1 | - | 1 | 566 | 6971949 | ||

XCR1
Genomic Map