siRNA Details
| Gene Symbol: Dusp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173745.5 | NP_776106.1 | 2 | 469 | ||
| GenBank mRNA | AK015917.1 | - | 2 | 453 | ||
| AK079664.1 | - | 2 | 465 | |||
| AK081916.1 | - | 2 | 428 | |||
| BC020036.1 | - | 2 | 463 | 3990751 | ||

Dusp18
Genomic Map