siRNA Details
| Gene Symbol: Zfp689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175163.3 | NP_780372.2 | 2 | 555 | ||
| GenBank mRNA | AK016830.1 | - | 2 | 391 | ||
| AK028417.1 | - | 2 | 518 | |||
| AK051667.1 | - | 2 | 369 | |||
| BC052084.1 | - | 2 | 555 | 6404620 | ||
| BU054587.1 | - | 2 | 555 | |||

Zfp689
Genomic Map