siRNA Details
| Gene Symbol: Aifm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291070.1 | NP_001277999.1 | 6 | 748 | ||
| NM_175178.4 | NP_780387.2 | 8 | 1154 | |||
| GenBank mRNA | AK012958.1 | - | 4 | 506 | ||
| AK049928.1 | - | 8 | 985 | |||
| AK158809.1 | - | 7 | 816 | |||
| BC046517.1 | - | 8 | 1140 | |||
| BC096476.1 | - | 7 | 929 | 4221224 | ||
| BC120685.1 | - | 7 | 783 | 40129608 | ||

Aifm3
Genomic Map