siRNA Details
| Gene Symbol: Tmem161b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175187.5 | NP_780396.2 | 10 | 1069 | ||
| GenBank mRNA | AK013305.1 | - | 9 | 898 | ||
| AK088393.1 | - | 10 | 1041 | |||
| BC037611.1 | - | 8 | 849 | |||
| BC051663.1 | - | 10 | 1017 | 30020101 | ||
| BC060067.1 | - | 10 | 1111 | |||

Tmem161b
Genomic Map