siRNA Details
| Gene Symbol: GPR15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005290.3 | NP_005281.1 | 1 | 465 | ||
| GenBank mRNA | AK313375.1 | - | 1 | 465 | ||
| BC069437.1 | - | 1 | 330 | 7262234 | ||
| BC101779.1 | - | 1 | 356 | 8069285 | ||
| BC101781.1 | - | 1 | 353 | 8069287 | ||
More Information
Set Membership: |
|

GPR15
Genomic Map