siRNA Details
| Gene Symbol: Hspa12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175199.3 | NP_780408.1 | 3 | 334 | ||
| GenBank mRNA | AB093239.1 | - | 3 | 278 | ||
| AK006867.1 | - | 3 | 318 | |||
| AK038491.1 | - | 3 | 306 | |||
| AK087732.1 | - | 4 | 491 | |||
| AK141925.1 | - | 3 | 335 | |||
| AK165073.1 | - | 3 | 362 | |||
| AY196789.1 | - | 3 | 333 | |||
| BC030362.1 | - | 3 | 304 | 5369051 | ||

Hspa12a
Genomic Map