siRNA Details
| Gene Symbol: Psmb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175204.4 | NP_780413.1 | 1 | 1478 | ||
| GenBank mRNA | AK037368.1 | - | 1 | 1476 | ||
| AK041393.1 | - | 1 | 1476 | |||
| AK162727.1 | - | 1 | 1480 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmb11| Gene Symbol: Psmb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175204.4 | NP_780413.1 | 1 | 1478 | ||
| GenBank mRNA | AK037368.1 | - | 1 | 1476 | ||
| AK041393.1 | - | 1 | 1476 | |||
| AK162727.1 | - | 1 | 1480 | |||