siRNA Details
| Gene Symbol: Ankrd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175207.4 | NP_780416.2 | 3 | 1185 | ||
| GenBank mRNA | AK010867.1 | - | 3 | 1350 | ||
| AK032641.1 | - | 3 | 1186 | |||
| AK051108.1 | - | 3 | 1205 | |||
| AK172167.1 | - | 4 | 1267 | |||
| BC023687.1 | - | 3 | 1159 | 5343549 | ||

Ankrd9
Genomic Map