siRNA Details
| Gene Symbol: Lysmd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191051.1 | NP_001177980.1 | 3 | 1896 | ||
| NM_175215.4 | NP_780424.1 | 3 | 1559 | |||
| GenBank mRNA | AK033681.1 | - | 3 | 1496 | ||
| AK143241.1 | - | 3 | 1897 | |||
| AK172203.1 | - | 4 | 1659 | |||
| BC030310.1 | - | 4 | 2366 | |||
| BC051181.1 | - | 1 | 1344 | |||
| BC057459.1 | - | 3 | 1506 | 4504057 | ||
| BC094328.1 | - | 4 | 1909 | |||

Lysmd4
Genomic Map