siRNA Details
| Gene Symbol: Rnf139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175226.4 | NP_780435.1 | 2 | 312 | ||
| GenBank mRNA | AK028400.1 | - | 2 | 265 | ||
| AK029069.1 | - | 2 | 313 | |||
| AK033506.1 | - | 2 | 246 | |||
| BC052901.1 | - | 2 | 261 | 30070643 | ||

Rnf139
Genomic Map