siRNA Details
| Gene Symbol: Tmem198b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178066.2 | NP_835167.1 | 5 | 2189 | ||
| GenBank mRNA | AK054166.1 | - | 3 | 3070 | ||
| BC037624.1 | - | 5 | 2189 | 4507848 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem198b| Gene Symbol: Tmem198b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178066.2 | NP_835167.1 | 5 | 2189 | ||
| GenBank mRNA | AK054166.1 | - | 3 | 3070 | ||
| BC037624.1 | - | 5 | 2189 | 4507848 | ||