siRNA Details
| Gene Symbol: Flrt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172160.1 | NP_001165631.1 | 3 | 1386 | ||
| NM_178382.4 | NP_848469.1 | 3 | 833 | |||
| GenBank mRNA | AK028252.1 | - | 3 | 827 | ||
| AK031464.1 | - | 3 | 827 | |||
| AK129369.1 | - | 3 | 1310 | |||
| AK161100.1 | - | 3 | 827 | |||
| AY495670.1 | - | 1 | 405 | |||
| BC052043.1 | - | 3 | 808 | 5702881 | ||

Flrt3
Genomic Map