siRNA Details
| Gene Symbol: Fam53a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178390.3 | NP_848477.2 | 6 | 1884 | ||
| GenBank mRNA | AK010537.1 | - | 6 | 1820 | ||
| AK017321.1 | - | 5 | 2229 | |||
| AK155077.1 | - | 7 | 1997 | |||
| AK163326.1 | - | 6 | 1856 | |||
| AK179669.1 | - | 1 | 285 | |||
| AK195761.1 | - | 1 | 271 | |||
| AK220276.1 | - | 4 | 8103 | |||
| BC016596.1 | - | 4 | 1309 | 3257498 | ||
| BC028656.1 | - | 6 | 1604 | 1244982 | ||

Fam53a
Genomic Map