siRNA Details
| Gene Symbol: Wipi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178398.4 | NP_848485.1 | 10 | 1174 | ||
| GenBank mRNA | AK010881.1 | - | 10 | 1200 | ||
| AK155113.1 | - | 10 | 1204 | |||
| AK169051.1 | - | 10 | 1148 | |||
| AK172030.1 | - | 10 | 1150 | |||
| BC044894.1 | - | 10 | 1199 | 4235237 | ||
| BC060262.1 | - | 1 | 25 | |||

Wipi2
Genomic Map