siRNA Details
| Gene Symbol: Urgcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077661.1 | NP_001071129.1 | 6 | 702 | ||
| NM_178623.3 | NP_848738.2 | 6 | 286 | |||
| GenBank mRNA | AK033577.1 | - | 6 | 260 | ||
| AK035580.1 | - | 6 | 698 | |||
| AK041953.1 | - | 7 | 951 | |||
| AK129378.1 | - | 5 | 344 | |||
| AK166495.1 | - | 5 | 186 | |||
| AK190812.1 | - | 3 | 75 | |||
| AK211902.1 | - | 2 | 53 | |||
| BC066994.1 | - | 6 | 583 | 5696000 | ||
| BY058620.1 | - | 6 | 286 | |||

Urgcp
Genomic Map