siRNA Details
| Gene Symbol: Urgcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077661.1 | NP_001071129.1 | 6 | 1381 | ||
| NM_178623.3 | NP_848738.2 | 6 | 965 | |||
| GenBank mRNA | AK033577.1 | - | 6 | 939 | ||
| AK035580.1 | - | 6 | 1377 | |||
| AK041953.1 | - | 7 | 1630 | |||
| AK129378.1 | - | 5 | 1023 | |||
| AK166495.1 | - | 5 | 865 | |||
| BC066994.1 | - | 6 | 1262 | 5696000 | ||

Urgcp
Genomic Map