siRNA Details
| Gene Symbol: Fbxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178624.6 | NP_848739.1 | 15 | 1608 | ||
| GenBank mRNA | AK013142.1 | - | 16 | 1744 | ||
| AK039010.1 | - | 15 | 1608 | |||
| AK044693.1 | - | 15 | 1605 | |||
| AK045742.1 | - | 15 | 1595 | |||
| AK141177.1 | - | 15 | 1598 | |||
| AK163472.1 | - | 15 | 1604 | |||
| BC096582.1 | - | 15 | 1594 | 3963599 | ||
| BC145665.1 | - | 15 | 1588 | 40130968 | ||
| BC145998.1 | - | 15 | 1588 | 40131301 | ||

Fbxl2
Genomic Map