siRNA Details
| Gene Symbol: Reep4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_180588.2 | NP_850919.1 | 8 | 1619 | ||
| GenBank mRNA | AK012301.1 | - | 8 | 1516 | ||
| BC033929.1 | - | 8 | 1247 | 1363539 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Reep4| Gene Symbol: Reep4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_180588.2 | NP_850919.1 | 8 | 1619 | ||
| GenBank mRNA | AK012301.1 | - | 8 | 1516 | ||
| BC033929.1 | - | 8 | 1247 | 1363539 | ||