siRNA Details
| Gene Symbol: 1700067K01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183097.2 | NP_898920.2 | 4 | 618 | ||
| GenBank mRNA | AK006917.1 | - | 4 | 887 | ||
| BC147276.1 | - | 4 | 624 | 9056316 | ||
| BC147277.1 | - | 4 | 624 | 9056320 | ||
| BC172025.1 | - | 4 | 621 | 9054311 | ||
| BC172026.1 | - | 4 | 621 | |||

1700067K01Rik
Genomic Map