siRNA Details
| Gene Symbol: 1700067K01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183097.2 | NP_898920.2 | 6 | 809 | ||
| GenBank mRNA | AK006917.1 | - | 6 | 1078 | ||
| BC147276.1 | - | 6 | 815 | 9056316 | ||
| BC147277.1 | - | 6 | 815 | 9056320 | ||
| BC172025.1 | - | 6 | 812 | 9054311 | ||
| BC172026.1 | - | 6 | 876 | |||

1700067K01Rik
Genomic Map