siRNA Details
Gene Symbol: Prss46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183103.2 | NP_898926.2 | 3 | 523 | ||
GenBank mRNA | AK007173.1 | - | 3 | 522 | ||
BC087878.1 | - | 2 | 480 | 30284660 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prss46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183103.2 | NP_898926.2 | 3 | 523 | ||
GenBank mRNA | AK007173.1 | - | 3 | 522 | ||
BC087878.1 | - | 2 | 480 | 30284660 |