siRNA Details
| Gene Symbol: Prss46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183103.2 | NP_898926.2 | 3 | 528 | ||
| GenBank mRNA | AK007173.1 | - | 3 | 527 | ||
| BC087878.1 | - | 2 | 485 | 30284660 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss46| Gene Symbol: Prss46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183103.2 | NP_898926.2 | 3 | 528 | ||
| GenBank mRNA | AK007173.1 | - | 3 | 527 | ||
| BC087878.1 | - | 2 | 485 | 30284660 | ||