siRNA Details
| Gene Symbol: 4931429L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183104.2 | NP_898927.2 | 10 | 1432 | ||
| GenBank mRNA | AK016488.1 | - | 10 | 1429 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4931429L15Rik| Gene Symbol: 4931429L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183104.2 | NP_898927.2 | 10 | 1432 | ||
| GenBank mRNA | AK016488.1 | - | 10 | 1429 | ||