siRNA Details
Gene Symbol: 4931429L15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183104.2 | NP_898927.2 | 9 | 1252 | ||
GenBank mRNA | AK016488.1 | - | 9 | 1249 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4931429L15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183104.2 | NP_898927.2 | 9 | 1252 | ||
GenBank mRNA | AK016488.1 | - | 9 | 1249 |