siRNA Details
| Gene Symbol: Iba57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270791.1 | NP_001257720.1 | 2 | 752 | ||
| GenBank mRNA | AK016031.1 | - | 2 | 686 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_183110.1 | - | 686 | Chr. - - [-] on Build | ||

Iba57
Genomic Map