siRNA Details
| Gene Symbol: 4932414N04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183113.3 | NP_898936.1 | 16 | 1389 | ||
| GenBank mRNA | AK016523.1 | - | 16 | 1390 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4932414N04Rik| Gene Symbol: 4932414N04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183113.3 | NP_898936.1 | 16 | 1389 | ||
| GenBank mRNA | AK016523.1 | - | 16 | 1390 | ||