siRNA Details
| Gene Symbol: Ccdc125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168386.2 | NP_001161858.1 | 9 | 1243 | ||
| NM_183115.5 | NP_898938.4 | 10 | 1321 | |||
| GenBank mRNA | AK017973.1 | - | 9 | 1006 | ||
| BC085249.1 | - | 10 | 1039 | 30726150 | ||

Ccdc125
Genomic Map