siRNA Details
| Gene Symbol: Naif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194335.2 | NP_919316.1 | 2 | 2638 | ||
| GenBank mRNA | AK029812.1 | - | 2 | 2639 | ||
| BC057613.1 | - | 894 | 6401522 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Naif1| Gene Symbol: Naif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194335.2 | NP_919316.1 | 2 | 2638 | ||
| GenBank mRNA | AK029812.1 | - | 2 | 2639 | ||
| BC057613.1 | - | 894 | 6401522 | |||