siRNA Details
| Gene Symbol: Dcaf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165980.1 | NP_001159452.1 | 2 | 430 | ||
| NM_001165981.1 | NP_001159453.1 | 2 | 430 | |||
| NM_001165982.1 | NP_001159454.1 | 2 | 430 | |||
| NM_198005.2 | NP_932122.2 | 2 | 430 | |||
| GenBank mRNA | AK014717.1 | - | 2 | 394 | ||
| AK032671.1 | - | 2 | 392 | |||
| AK079462.1 | - | 2 | 224 | |||
| AK081839.1 | - | 2 | 429 | |||
| AK160679.1 | - | 2 | 397 | |||
| BC031546.1 | - | 2 | 322 | 3669736 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001177688, XM_006234347, BC160919.

Dcaf17
Genomic Map