siRNA Details
| Gene Symbol: Fam160a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242363.2 | NP_001229292.1 | 4 | 987 | ||
| NM_001242364.2 | NP_001229293.1 | 3 | 896 | |||
| NM_001242365.2 | NP_001229294.1 | 3 | 896 | |||
| NM_199009.3 | NP_950174.1 | 3 | 896 | |||
| GenBank mRNA | AK085350.1 | - | 3 | 846 | ||
| AK122552.1 | - | 3 | 824 | |||
| AK154173.1 | - | 4 | 931 | |||
| AK155273.1 | - | 4 | 932 | |||
| AK159716.1 | - | 3 | 851 | |||
| AK169996.1 | - | 4 | 965 | |||
| BC018464.1 | - | 3 | 826 | |||
| BC021532.1 | - | 1 | 223 | |||
| BC043717.1 | - | 2 | 412 | |||
| BC063093.1 | - | 3 | 853 | 6332038 | ||

Fam160a2
Genomic Map