siRNA Details
| Gene Symbol: Thap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199042.2 | NP_950243.1 | 3 | 614 | ||
| GenBank mRNA | BC038639.1 | - | 3 | 570 | 5118383 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thap1| Gene Symbol: Thap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199042.2 | NP_950243.1 | 3 | 614 | ||
| GenBank mRNA | BC038639.1 | - | 3 | 570 | 5118383 | |